Learn More
Medchemexpress LLC Allantoin (Standard) | 97-59-6 | MFCD00005260 | 158.12 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYN0543R
Encompass_Preferred
Allantoin (Standard) is the analytical standard of Allantoin, intended for research and analytical applications. It acts as a skin conditioning agent, promoting healthy skin and stimulating new, healthy tissue growth. This compound is typically used as an analytical standard for assays and in qualitative, quantitative, and methodological research experiments.
  • Promotes healthy skin
  • Stimulates new and healthy tissue growth
  • Used in HPLC, GC, and MS research experiments
  • Reference for assays
Catalog No. 50-003-89325