Learn More
Cayman Chemical allcis14 17 20 23 26 29Dotria
SDP
Supplier:  Cayman Chemical 10562500 UG
Encompass_Preferred
A very long-chain fatty alcohol; an intermediate in the synthesis of all-cis-14,17,20,23,26,29-dotriacontahexaenoic acid
Catalog No. 50-288-9254