Learn More
Grainger ALLEGRO Confined Space Fan, 115 V AC
Supplier:  Grainger 5GVX4
Encompass_Preferred
The Allegro Confined Space Fan is a 115 V AC, 1/3 HP fan designed for ventilating confined spaces. It features a lightweight aluminum construction with a GFCI plug for safety. The fan provides 5000 CFM airflow and 12" blade diameter. It has a duct diameter of 12" and includes 25' of ducting. The fan's motor is totally enclosed for hazardous locations. It comes with a carrying handle and is CSA approved. The fan weighs 43 lbs and measures 14" W x 14" D x 15.5" H.
Catalog No. NC2602730