Learn More
Medchemexpress LLC HY-N0315 50mg Medchemexpress, Allicin CAS:539-86-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN031550MG
Encompass_Preferred
Medchemexpress, HY-N0315 50mg Allicin CAS:539-86-6 Allicin (diallyl thiosulfinate), a highly potent natural antimicrobial activity substance, inhibits growth of a variety of microorganisms, among them antibiotic-resistant strains [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3515