Learn More
Medchemexpress LLC Alliin | 556-27-4 | 177.22 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN066110MM/1ML
Encompass_Preferred
Alliin is an orally active sulfoxide compound that can be isolated from garlic. It exhibits hypoglycemic, antioxidant, anti-inflammatory, and antitumor activities.
  • Orally active compound.
  • Can be isolated from garlic.
  • Exhibits hypoglycemic activity.
  • Exhibits antioxidant activity.
  • Exhibits anti-inflammatory activity.
  • Exhibits antitumor activity.
Catalog No. 50-003-88470