Learn More
Medchemexpress LLC ALLO-2 | 1357350-60-7 | 99.7% | 371.32 g/mol | C18H12F3N5O | 1 ML
SDP
Supplier:  Medchemexpress LLC HY11740710MM 1ML
Encompass_Preferred
ALLO-2 is a small-molecule Smoothened (Smo) mutant antagonist that inhibits Smo agonist Hh-Ag1.5-induced luciferase expression in TM3-Gli-Luc cells with an IC50 of 6 nM. It is supplied for research use as solid material or as a pre-made 10 mM solution in DMSO (1 mL).
  • Potent Smo mutant antagonist with IC50 ≈ 6 nM.
  • Active against drug-resistant Smoothened mutants, suitable for pathway inhibition studies.
  • Available as solid or as a pre-made 10 mM solution in DMSO for rapid in vitro use.
  • High purity (99.7%), appropriate for research applications.
  • Useful for luciferase reporter assays and other cell-based Smo activity assays.
Catalog No. 50-002-26663