Learn More
Medchemexpress LLC Allo-2 | 1357350-60-7 | 99.7% | 371.32 g/mol | C18H12F3N5O | 50 MG
SDP
Supplier:  Medchemexpress LLC HY11740750MG
Encompass_Preferred
ALLO-2 is a potent, drug-resistant Smoothened (Smo) mutant antagonist that inhibits Hh-Ag1.5-induced luciferase expression in TM3-Gli-Luc cells with a reported IC50 of 6 nM. The compound is provided as a high-purity, research-grade small molecule (CAS 1357350-60-7; C18H12F3N5O; MW 371.32).
  • Potent Smo mutant antagonist with IC50 ≈ 6 nM
  • High reported chemical purity (99.72%)
  • Characterized molecular formula and molecular weight for reproducibility
  • Intended for non-clinical research applications and in vitro assays
Catalog No. 50-002-28643