Learn More
Cayman Chemical Allopurinol Inhibitors
SDP
Supplier:  Cayman Chemical 10012597100 G
Encompass_Preferred
An isomer of hypoxanthine that inhibits xanthine oxidoreductase (IC50 values between 0.2 and 50 μM); lowers serum and urinary uric acid levels in vivo and is used to treat gout and hyperuricemia
Catalog No. 50-289-0223