Learn More
Selleck Chemical LLC Allopurinol riboside E4922-100MG
SDP
Supplier:  Selleck Chemical LLC E4922100MG
Encompass_Preferred
Allopurinol riboside a metabolite of allopurinol competitively inhibits the action of purine nucleoside phosphorylase on inosine in vitro with a Ki value of 277 M It displays potent activities against parasites
Catalog No. 50-000-41571