Learn More
Medchemexpress LLC Allosecurinine | 884-68-4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN23775MG
Encompass_Preferred
Allosecurinine (Phyllochrysine) is a Securinega alkaloid isolated from *Phyllanthus glaucus*. Securinega alkaloids are natural products found in some *Phyllanthaceae* species and exhibit important biological activities, including stimulating the Central Nervous System (CNS), and possessing antitumor, anti-malarial, and antibacterial properties.
  • Securinega alkaloid
  • Isolated from *Phyllanthus glaucus*
  • Exhibits stimulating effects on the Central Nervous System (CNS)
  • Demonstrates antitumor activity
  • Shows anti-malarial activity
  • Possesses antibacterial activity
Catalog No. 50-004-18411