Learn More
Medchemexpress LLC Alloxazine | 490-59-5 | 100MG
SDP
Supplier:  Medchemexpress LLC HY123085100MG
Encompass_Preferred
Alloxazine is a selective A2b antagonist Alloxazine completely block 5 N-Ethylcarboxamido adenosine (NECA)-mediated cyclic AMP accumulation with an IC50 of 2 9 M Alloxazine can be used for the research of cancer[1][2]
Catalog No. 50-002-37937