Learn More
Mac-Mod Avantor Hichrom Organic Acid HPLC Columns
SDP
Supplier:  Mac-Mod 9046
Encompass
Avantor Hichrom Organic Acid HPLC columns are ion exclusion columns packed with sulphonated polystyrene-divinylbenzene
Avantor Hichrom Organic Acid HPLC columns are available in 4 versions
oOA-1000 and OA-2000. These columns exhibit excellent selectivity for aliphatic and aromatic acids. Note: As with most ion exclusion columns, a column heater in necessary for normal operating procedures
oIOA-1000 and IOA-2000 columns are suitable for the separation of citric and other acids from glucose and fructose
Avantor Hichrom Organic Acid HPLC columns offer rapid analysis of organic acids and alcohols
Avantor Hichrom Organic Acid HPLC columns are built on pH stable polymer resin
Avantor Hichrom Organic Acid HPLC columns operate in 100% aqueous isocratic mobile phases only, no organic solvents can be used
Avantor Hichrom Organic Acid are classified by the USP as L17
Catalog No. NC1576257