Learn More
Medchemexpress LLC Allyl alpha-D-galactopyranoside | 48149-72-0 | MFCD01320363 | ≥98.0% | 220.22 g/mol | C9H16O6 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYW039915
Encompass_Preferred
Allyl α-D-galactopyranoside is an allyl glycoside of D-galactose used as a synthetic intermediate and biochemical reagent in glycobiology and carbohydrate chemistry. It is supplied as a solid (C9H16O6; 220.22 g/mol) and is commonly used in glycan synthesis, enzymology studies, and investigations of carbohydrate-protein interactions.
  • Useful synthetic intermediate for oligosaccharide and glycan synthesis.
  • Suitable for enzymatic studies of glycosidases and glycosyltransferases.
  • High purity (≥98.0%) supports reproducible experimental results.
  • Solid form facilitates handling and storage.
  • Soluble in common organic solvents for synthesis and analysis.
Catalog No. 50-003-03541