Learn More
Strem, An Ascensus Company CAS# 478980-01-7. 250mg. Allylchloro[1,3-bis(2,6-di-i-propylphenyl)-4,5-dihydroimidazol-2-ylidene]palladium(II), 97%. MFCD07782017
SDP
Supplier:  Strem, An Ascensus Company 460039250MG
Encompass
CAS# 478980-01-7. 250mg. Allylchloro[1,3-bis(2,6-di-i-propylphenyl)-4,5-dihydroimidazol-2-ylidene]palladium(II), 97%. MFCD07782017. Molecular Weight: 573.55. Molecular Formula: C30H43ClN2Pd. Color/form: white pwdr. Strem# 46-0039. http://www.strem.com/catalog/v/46-0039/
Catalog No. 50-901-12275