Learn More
Medchemexpress LLC ((Allyloxy)carbonyl)-L-valyl-L-alanine | 330970-70-2 | MFCD28387723 | >95.0% | 272.30 g/mol | C12H20N2O5 | 25 G

Supplier: Medchemexpress LLC 25GHYW076556
Alloc-Val-Ala-OH (((Allyloxy)carbonyl)-L-valyl-L-alanine) is an Alloc-protected dipeptide used as a synthetic building block in medicinal chemistry and in linker/payload synthesis for antibody-drug conjugates. It is susceptible to enzymatic cleavage by cathepsin B and is supplied for research use.
- Cas number 330970-70-2.
- Molecular formula C12H20N2O5; molecular weight 272.30 g/mol.
- Purity >95.0%.
- Available in multiple pack sizes, including 25 G.
- Alloc-protected N-terminal enables selective deprotection.
- Useful as a building block in linker and payload synthesis for ADCs.
- For research use only; consult SDS and COA for handling and safety.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.