Learn More
Medchemexpress LLC ((Allyloxy)carbonyl)-L-valyl-L-alanine | 330970-70-2 | MFCD28387723 | ≥95.0% | 272.30 g/mol | C12H20N2O5 | 250 MG

Supplier: Medchemexpress LLC 250MGHYW076556
Alloc-Val-Ala-OH is the N-allyloxycarbonyl (Alloc) protected dipeptide ((Allyloxy)carbonyl)-L-valyl-L-alanine used as a protected building block in peptide synthesis and organic synthesis. It is supplied as a white to off-white solid with high purity suitable for peptide coupling, ADC linker assembly, and related synthetic applications.
- White to off-white solid
- Purity ≥95.0%
- Molecular weight 272.30 g/mol
- Molecular formula C12H20N2O5
- Available in 250 MG and larger pack sizes
- Recommended storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.