Learn More
Medchemexpress LLC Allylthiourea | 109-57-9 | 100.0% | 50 G
SDP
Supplier:  Medchemexpress LLC HYB054350G
Encompass_Preferred
Allylthiourea selectively inhibits ammonia oxidation, commonly used to suppress nitrification by targeting ammonia monooxygenase and chelating copper. It also demonstrates anticancer activity, exhibiting cytotoxicity against the MCF-7 cell line. This compound is valuable for research in micropollutant biodegradability and cancer studies.
  • Selectively inhibits ammonia oxidizers.
  • Targets ammonia monooxygenase to inhibit nitrification.
  • Chelates copper in the active site, suppressing enzyme activity.
  • Exhibits cytotoxicity against the MCF-7 cell line (IC50 of 5.22 mM).
  • Can be used in micropollutant biodegradability research.
  • Applicable in cancer studies.
  • Decreases intracellular copper in Methylosinus trichosporium OB3b, allowing sMMO production.
Catalog No. 50-004-07585