Learn More
Medchemexpress LLC Allylthiourea (Thiosinamine) | 109-57-9 | 99.95% | 100 G
SDP
Supplier:  Medchemexpress LLC HYB0543100G
Encompass_Preferred
Allylthiourea can selectively inhibit the oxidation of ammonia. It is commonly used to inhibit nitrification by targeting ammonia monooxygenase and chelating copper in the active site to suppress its activity. Allylthiourea also exhibits anticancer activity, showing cytotoxicity against the MCF-7 cell line with an IC50 of 5.22 mM. It can be utilized in research related to micropollutant biodegradability and cancer studies.
  • Selectively inhibits ammonia oxidizers.
  • Decreases intracellular copper in *Methylosinus trichosporium* OB3b.
  • Used to inhibit nitrification by targeting ammonia monooxygenase.
  • Exhibits cytotoxicity against MCF-7 cell line.
  • Useful for micropollutant biodegradability and cancer studies.
Catalog No. 50-004-07624