Learn More
Medchemexpress LLC Allyltrimethylsilane | 762-72-1 | MFCD00008635 | 99.6% | 114.26 g/mol | C6H14Si | 100 G
SDP
Supplier:  Medchemexpress LLC HYW019828100G
Encompass_Preferred
Allyltrimethylsilane is an organosilicon reagent (CAS 762-72-1) used in organic synthesis as an allyl transfer agent and building block. It is a colorless to light yellow liquid with formula C6H14Si and a molecular weight of 114.26 g/mol, commonly employed in allylation and silicon-based transformations at laboratory scale.
  • High purity (99.57%) suitable for synthetic applications.
  • Useful as an allyl transfer reagent in carbon-carbon bond formation.
  • Compatible with common Lewis acid-catalyzed reactions.
  • Liquid form allows easy dosing and handling in reactions.
  • Available supporting documents such as SDS and COA for technical and safety data.
Catalog No. 50-003-01855