Learn More
Medchemexpress LLC Almotriptan (malate) | 181183-52-8 | 1 ML
Supplier: Medchemexpress LLC HYB038310MM/1ML
Almotriptan malate is an orally active, highly selective agonist of the 5-HT1B/1D receptor, with potential applications in migraine research. It is suitable for use as a laboratory chemical in the manufacture of substances.
- Highly selective 5-HT1B/1D receptor agonist
- Induces intracranial vasoconstriction
- Inhibits nociceptive neurotransmission in the trigeminocervical complex
- Suppresses release of vasoactive peptides from trigeminal nerve endings
- Solid physical state
- Melting point 170-172°C