Learn More
Cayman Chemical AlolIthocholIc AcId 10mg
SDP
Supplier:  Cayman Chemical 3815810 MG
Encompass_Preferred
A secondary allomonohydroxy bile acid; activates BK/KCa channels (EC50 = 44.21 µM in Xenopus oocytes expressing rat channels); fecal levels are increased in patients with colorectal cancer; has been found in the urine of infants with biliary atresia
Catalog No. 50-291-5859