Learn More
TARGETMOL CHEMICALS INC Alosetron hydrochloride 50MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T252550MG
Encompass_Preferred
Also available in 10 mg, 25 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Alosetron hydrochloride (GR 68755) is the hydrochloride salt form of alosetron, a potent and selective 5-HT3 receptor antagonist. Alosetron blocks the actions of serotonin at 5-HT3 sites in the peripheral nervous system, particularly on enteric and nociceptive sensory neurons, thereby affecting the regulation of visceral pain, decreasing gastrointestinal contraction and motility, and decreasing gastrointestinal secretions. Purity 100%
Catalog No. 50-287-7606