Learn More
TARGETMOL CHEMICALS INC ALOX15-IN-2 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T6260610MG
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ALOX15-IN-2 is a potent inhibitor of ALOX15 linoleate oxygenase activity inhibiting the oxidation of linoleic acid (LA) and arachidonic acid (AA) the immediate homologs of ALOX15 with IC50s of 1.55 and 2.79 uM respectively. purity: 99%
Catalog No. 50-257-9467