Learn More
Selleck Chemical LLC Aloxistatin
SDP
Supplier:  Selleck Chemical LLC S739325MG
Encompass_Preferred
Aloxistatin (E64d) is an irreversible and membrane-permeable cysteine protease inhibitor with blood platelet aggregation inhibiting activity The cysteine protease cathepsin L is required for SARS-CoV-2 viral entry and aloxistatin treatment reduced cellular entry of SARS-CoV-2 pseudovirions by 92 3%
Catalog No. 50-000-01528