Learn More
Selleck Chemical LLC Aloxistatin (E64d) 5mg 88321-09-9 E-64d
SDP
Supplier:  Selleck Chemical LLC S73935MG
Encompass_Preferred
Aloxistatin (E64d) is an irreversible and membrane-permeable cysteine protease inhibitor with blood platelet aggregation inhibiting activity. The cysteine protease cathepsin L is required for SARS-CoV-2 viral entry, and aloxistatin treatment reduced cellular entry of SARS-CoV-2 pseudovirions by 92.3%. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4733