Learn More
Beckman Coulter 1x1 Tip Load Adapter Plate (ALP) for Biomek workstations - Beckman Coulter C02751
Supplier:  Beckman Coulter C02751
Encompass
Beckman Coulter C02751 1x1 Tip Load Adapter Plate (ALP) is an accessory designed to mount on the deck of Biomek automated workstations to enable 1x1 tip loading and other application-specific functions. It is part of the Automated Labware Positioners (ALP) family and supports workflows that include tip washing, shaking, heating/cooling, and reagent handling.
  • Adapter plate for 1x1 tip loading.
  • Integrates onto the deck of compatible automated workstations.
  • Supports application-specific tasks such as tip washing and shaking.
  • Part of a modular automated labware positioner family for workflow automation.
  • Package quantity and physical dimensions are not specified on the product page.
Catalog No. NC1977133