Learn More
Medchemexpress LLC Alp/carbonic anhydrase-in-1 | 2091887-74-8 | 99.9% | 224.30 | C15H16N2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY15713910MG
Encompass_Preferred
ALP/Carbonic anhydrase-IN-1 is a research reagent: a small molecule dual inhibitor of carbonic anhydrase isoforms and alkaline phosphatase intended for in vitro biochemical and cell-based studies. It is provided as a high-purity solid suitable for screening and assay development.
  • Dual inhibitor of carbonic anhydrase and alkaline phosphatase.
  • High purity (99.9%) for reproducible experimental results.
  • Molecular weight 224.30 and formula C15H16N2.
  • Supplied as a 10 MG research pack for laboratory use.
  • Documented inhibitory activity against multiple CA isoforms and ALP, useful for mechanism and screening studies.
Catalog No. 50-002-87881