Learn More
Medchemexpress LLC Alpelisib (hydrochloride) | 1584128-91-5 | C19H23ClF3N5O2S | 200 MG
SDP
Supplier:  Medchemexpress LLC HY15244A200MG
Encompass_Preferred
Alpelisib hydrochloride is a potent, orally active, and selective PI3Kα inhibitor. It demonstrates antineoplastic activity and effectively inhibits common PIK3CA somatic mutations.
  • Potent, orally active, and selective PI3Kα inhibitor
  • Effectively inhibits common PIK3CA somatic mutations
  • Potently inhibits Akt phosphorylation in PI3Kα-transformed cells
  • Induces G0/G1 phase cell cycle arrest in osteosarcoma cells
  • Significantly reduces tumor volumes in osteosarcoma animal models
Catalog No. 50-003-72414