Learn More
Medchemexpress LLC Alpha-1,4-Galactosyltransferase (LgtC) | 52725-57-2 | 32 kDa | 5 U

Supplier: Medchemexpress LLC HYE700475U
alpha-1,4-Galactosyltransferase (LgtC) (A4GALT) is a glycosphingolipid-specific glycosyltransferase. It transfers a galactose to the alpha-1,4 position of lactosylceramide to form globotriaosylceramide, and can be utilized for the synthesis of P1 blood group antigens.
- Recombinantly expressed in *E. coli* with an N-terminal His-tag
- Molecular weight of 32 kDa
- Buffer composition: 20 mM Tris-HCl, pH 7.5, 5% glycerol
- Transfers galactose to sugar chains with Gal as the non-reducing end
- Enzyme activity: ≥1 U/mg solid
- Unit definition: One unit is defined as the amount of enzyme that will convert 1 μmol Gal from UDP-Gal to lactose, forming Galα1,4 lac per minute at 37°C
- Appearance: Liquid, colorless to light yellow
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.