Learn More
Alpco Ltd Alpha 1-Acid Glycoprotein ELISA
SDP
Supplier:  Alpco Ltd 41AGPHUE01
Encompass
The Alpha 1-Acid Glycoprotein ELISA is a highly sensitive two-site enzyme linked immunoassay for measuring AGP in the biological fluids of humans.
Catalog No. NC2885134