Learn More
CPC Scientific H-Asn-Gln-Glu-Gln-Val-Ser-Pro-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific THRO002A
Encompass
This peptide is the functional factor XIIIa substrate. Alpha-2-plasmin inhibitor, also termed primary plasmin inhibitor or antiplasmin, is the most potent and rapidly acting of the plasmin inhibitors and is thought to be important in the regulation of fibrinolysis in vivo. It has been used in assays to develop the potential of fibrin to promote nerve regeneration by enzymatically incorporating exogenous neurite-promoting heparin-binding peptides.SEQUENCE: H-Asn-Gln-Glu-Gln-Val-Ser-Pro-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: NQEQVSPMOLECULAR FORMULA: C32H52N10O14MOLECULAR WEIGHT: 800.82STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: SYNONYMS: RESEARCH AREA: HematologyREFERENCES:Lloyd, A. et al. Pharm. Sci. Techn.Today 3, 257 (2000)Sakiyama, S. et al. FASEB J. 13, 2214 (1999)Holmes, W. et al. J. Biol. Chem. 262, 1659 (1987)SKU(s): THRO-002A
Catalog No. 50-211-0563