Learn More
Sigma Aldrich Fine Chemicals Biosciences alpha,alpha-Dichlorotoluene 95% | 98-87-3 | MFCD00000829 | 25G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 25104625G
Encompass
alpha,alpha-Dichlorotoluene 95% | Purity: 95% | Mol Wt: 161.03 | 98-87-3 | MFCD00000829 | 25G
Catalog No. 50-185-9863