Learn More
Sigma Aldrich Fine Chemicals Biosciences alpha Amylase from human s
Supplier:  Sigma Aldrich Fine Chemicals Biosciences A0521500UN
Encompass
amylase is the main secretory protein constituent of saliva. It contains 496 amino acids and has two isoforms glycosylated and non-glycosylated. amylase gene is located on human chromosome 1p21.1. amylase is mainly produced in pancreas and saliva.
Catalog No. 50-290-3463