Learn More
CPC Scientific H-Ala-Pro-Arg-Leu-Arg-Phe-Tyr-Ser-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific BACP002A
Encompass
ONE-LETTER SEQUENCE: APRLRFYSMOLECULAR FORMULA: C47H72N14O11MOLECULAR WEIGHT:1009.19STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [87549-53-9], RESEARCH AREA: HormonalREFERENCES: B.S. Rothman et al.; PNAS, 80, 5753 (1983)
Catalog No. 50-210-8835