Learn More
CPC Scientific H-Arg-Tyr-Leu-Gly-Tyr-Leu-OH (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific ACAS001A
Encompass
The alpha-casein was the most potent opioid in opioid activity bioassays.SEQUENCE: H-Arg-Tyr-Leu-Gly-Tyr-Leu-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: RYLGYLMOLECULAR FORMULA: C38H57N9O9MOLECULAR WEIGHT: 783.93STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [83471-50-5]SYNONYMS: -Casein (90-95)RESEARCH AREA: PainREFERENCES:S.Loukas et al., Biochemistry, 22, 4567 (1983)SKU(s): ACAS-001A
Catalog No. 50-210-8571