Learn More
Medchemexpress LLC Alpha-crystallin A c 2ug | 2ug
SDP
Supplier:  Medchemexpress LLC HYP78722UG
Encompass_Preferred
Alpha-crystallin A chain/CRYAA Protein Human (His) is a recombinant Alpha-crystallin A chain protein with a His tag Alpha-crystallin A chain/CRYAA is a small heat shock protein and molecular chaperone that prevents nonspecific aggregation of denaturing proteins Several point mutations in the alphaA-crystallin gene cause congenital human cataracts by unknown mechanisms[1]
Catalog No. 50-001-95092