Learn More
Abcam alpha-Cyclode x trin, non-reducing cyclic saccharide, 25G
Supplier:  Abcam AB14627225G
Encompass_Preferred
MW 972.86 g/mol. Non-reducing cyclic saccharide consisting of six α-1,4-linked D-glucopyranosyl units produced by the action of cyclodextrin glucosyltransferase (CGTase, EC 2.4.1.19) on hydrolyzed starch. Useful for the solubilization of nonpolar macromolecules such as fatty acids, lipids, and cholesterol for use in cell culture applications. Cyclodextrins are low molecular weight organic molecules which form water-soluble complexes with various compounds.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4770