Learn More
Medchemexpress LLC Alpha-Cypermethrin | 67375-30-8 | 99.7% | 200 MG

Supplier: Medchemexpress LLC HYB0874200MG
Alpha-Cypermethrin is an effective insecticide, demonstrated to be active against house flies. This compound is also being investigated in clinical trials for conditions such as Malaria, suggesting potential applications in vector control. It is presented as a white to off-white solid with a high purity of 99.66%.
- Purity: 99.66%
- Appearance: Solid
- Color: White to off-white
- Molecular weight: 416.30
- Formula: C22H19Cl2NO3
- Shipping conditions: Room temperature (in continental US; may vary elsewhere)
- Storage conditions for powder: -20°C for 3 years, 4°C for 2 years
- Storage conditions in solvent: -80°C for 6 months, -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.