Learn More
Medchemexpress LLC Alpha-Estradiol | 57-91-0 | MFCD00064144 | 99.5% | 1 G
SDP
Supplier:  Medchemexpress LLC HYB0141A1G
Encompass_Preferred
Alpha-Estradiol is a weak estrogen and a 5α-reductase inhibitor. It is utilized as a topical medication for the treatment of androgenic alopecia and is classified as a human endogenous metabolite.
  • Weak estrogen and 5 alpha-reductase inhibitor
  • Inhibits testosterone metabolism catalyzed by 5 alpha-reductase
  • Attenuates LPS-induced inflammatory markers in various cell types
  • Decreases NFκB-p65 and increases ER alpha protein expression
  • Reduces central avascular/total retina area in mouse models
  • Decreases malondialdehyde (MDA) levels in hyperoxia-exposed pups
  • Reduces NADPH-oxidase-positive cells, concentration, and activity in pup retinas
Catalog No. 50-004-50930