Learn More
AdipoGen alpha-GalCer Dansylated
SDP
Supplier:  AdipoGen AGCN20514C500
Encompass_Preferred
Chemical. CAS 424823-23-4. Formula C62H111N3O10S. MW 1090.6. Fluorescent alpha-GalCer derivative. Potent stimulator of invariant natural killer T iNKT cells similar to alpha-GalCer determined by CD86 expression in DCs and B cells, and IL-2 production by iNKT cells. Specific ligand of the lipid-binding MHC class I-like protein CD1d in human and mouse NKT cells. Used as a fluorescent marker for the uptake of the glycolipid by dendritic cells DCs in vitro and to better understand the mechanism of iNKT cell activation.
Catalog No. 50-256-2503