Learn More
Medchemexpress LLC alpha-GalNAc-TEG-N3 | 882873-70-3 | C14H26N4O8 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1518635MG
Encompass_Preferred
alpha-GalNAc-TEG-N3 is a click chemistry reagent designed for binding between nucleic acids, lipids, proteins, and other molecules. It contains an azide group, allowing it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules and ring strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing DBCO or BCN groups.
  • High yield
  • High specificity
  • Simplicity
  • Enables binding of nucleic acids, lipids, proteins, and other molecules
  • Contains an azide group for click chemistry reactions
  • Supports copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Supports ring strain-promoted alkyne-azide cycloaddition (SPAAC)
Catalog No. 50-003-73901