Learn More
Medchemexpress LLC Alpha-isomethyl ionone | 127-51-5 | MFCD00034582 | 65.9% | 206.32 g/mol | C14H22O | 5 G

Supplier: Medchemexpress LLC HYW1277405G
Alpha-isomethyl ionone (α-cetone) is a synthetic fragrance ingredient and small-molecule reagent (CAS 127-51-5) commonly used in perfume formulations and laboratory analyses. It is a clear, colorless to pale yellow liquid with molecular formula C14H22O and molecular weight 206.32 g/mol. The material is supplied in small quantities as an analytical standard with listed purity ≈65.9%.
- Used as a fragrance ingredient and analytical reagent.
- Clear, colorless to pale yellow liquid appearance.
- Molecular formula C14H22O; molecular weight 206.32 g/mol.
- CAS number 127-51-5 for unambiguous identification.
- Typical applications include perfume formulation and analytical standards for GC/LC.
- Supplied in small quantities suitable for research and analysis.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.