Learn More
Medchemexpress LLC Alpha-lactose monohydrate | 5989-81-1 | MFCD00150747 | >99.0% | 360.31 g/mol | C12H24O12 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYW08790410MM/1ML
Encompass_Preferred
Alpha-lactose monohydrate is a disaccharide derived from glucose and galactose commonly used as an excipient, diluent, and analytical standard in pharmaceutical and biochemical research. It is available either as a bulk solid or as small-volume prepared solutions for laboratory use.
  • Used as an excipient and diluent in pharmaceutical formulations.
  • Serves as an analytical reference standard and reagent for biochemical assays.
  • Available as solid bulk material and as small-volume solution formulations for convenience.
  • Chemical formula C12H24O12 and molecular weight 360.31 g/mol.
  • Supplied with product documentation such as datasheet, certificate of analysis, and safety data sheet.
Catalog No. 50-002-08312