Learn More
Medchemexpress LLC Alpha-linolenic acid | 463-40-1 | MFCD00065720 | 99.3% | 278.43 g/mol | C18H30O2 | 10 MG

Supplier: Medchemexpress LLC HYN0728R10MG
Alpha-linolenic acid analytical standard for research and analytical applications. This omega-3 essential fatty acid (CAS 463-40-1) has formula C18H30O2 and molecular weight 278.43 g/mol. Supplied as a 10 MG standard with HPLC purity 99.32%. Store at -20°C under nitrogen; soluble in DMSO (100 mg/mL).
- Provides a high-purity reference standard for analytical assays.
- Confirmed HPLC purity of 99.32% for accurate quantitation.
- Small 10 MG quantity suited for analytical labs and standards preparation.
- Soluble in DMSO at 100 mg/mL; multiple vehicle options for in vitro use.
- Recommended storage at -20°C under nitrogen to maintain stability.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.