Learn More
AdipoGen alpha-Mannosylceramide (1 mg)
SDP
Supplier:  AdipoGen AGCR13594M001
Encompass_Preferred
Chemical. CAS: 320610-63-7. Formula: C50H99NO9. MW: 858.3. Potent stimulator of natural killer T (NKT) cells expressing the invariant Valpha19-Jalpha26 T cell receptor alpha chain (Valpha19 NKT cells).
Catalog No. 50-205-3961