Learn More
Medchemexpress LLC Alpha-MCPG | 146669-29-6 | MFCD00210205 | 99.1% | 209.20 g·mol⁻¹ | C10H11NO4 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10037150MG
Encompass_Preferred
Alpha-MCPG is a research-grade competitive antagonist of group I and II metabotropic glutamate receptors (mGluRs). It is supplied as a white to off-white solid and is used in neuroscience studies to inhibit mGluR-mediated signaling and to investigate synaptic plasticity mechanisms such as theta-burst stimulation-induced shifts in hippocampal neurons.
  • Competitive antagonist of group I and II metabotropic glutamate receptors.
  • Useful for blocking theta-burst stimulation-induced synaptic changes in rodent hippocampal preparations.
  • White to off-white solid suitable for in vitro and ex vivo assays.
  • High purity: 99.1%.
  • Molecular weight 209.20 g·mol⁻¹; formula C10H11NO4; CAS 146669-29-6.
Catalog No. 50-002-12706