Learn More
ILEX LIFE SCIENCES LLC ALPHA MEM P04-21060 500 ML
SDP
Supplier:  ILEX LIFE SCIENCES LLC P0421060
Encompass_Preferred
PAN-Biotech Alpha MEM Eagle Cell Culture Medium, w: L-Glutamine, w/o: Ribonucleosides, w/o: Deoxyribonucleosides, w: 2.2 g/L NaHCO3 (500 ml). NUCLEOSIDES: Without nucleosides. BICARBONATE: With 2.2 g/L Sodium Bicarbonate. GLUCOSE: With glucose. GLUTAMINE: With L-glutamine. PHENOL RED: With Phenol Red. HEPES: Without HEPES. SODIUM PYRUVATE: With sodium pyruvate. STERILE: Yes. STORAGE TEMP: +2-8°C. MANUFACTURER: PAN-Biotech GmbH. COUNTRY OF ORIGIN: Germany. DESCRIPTION: Minimum Essential Medium Eagle - Alpha Modification is a different formulation of MEM Eagle with a higher concentration of amino acids, lipoic acid, vitamins, and pyruvate. MEM-alpha was developed primarily for cultivation of hamster kidney cells but is suitable for a broad range of mammalian cells. Among others, the alpha-MEM promotes the growth and progeny of bone marrow cells in suspension culture and monolayer. MEM-alpha is also used as a separation medium or for the out-breeding of amniotic cells.
Catalog No. 50-273-3814