Learn More
CPC Scientific Ac-Ser-Tyr-Ser-Met-Glu-His-Phe-Arg-Trp-Gly-Lys-Pro-Val-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific MSHS001A
Encompass
Peptide that stimulates skin darkening in amphibians and can be used for controlling learning, feeding and motivating in mammals. ONE-LETTER SEQUENCE: Ac-SYSMEHFRWGKPV-NH2MOLECULAR FORMULA: C77H109N21O19S1MOLECULAR WEIGHT:1664.91STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [581-05-5], SYNONYMS: melanotropins, acetyl-ACTH (1-13) amide, -melanotropin (human), α-MSHRESEARCH AREA: HormonalREFERENCES: J.I. Harris et al., Nature, 179, 1346 (1957)
Catalog No. 50-210-9835