Learn More
Santa Cruz Biotechnology ALPHA TUBULIN B-7
Supplier:  Santa Cruz Biotechnology SC5286
Encompass
alpha Tubulin (B-7), Isotype: Mouse IgG2a, Species: Mouse, Rat, Human, Epitope: 149-448 (h), MW: 54 kd, Applications: WB, IP, IHC, FC.
Catalog No. NC9792938