Learn More
Medchemexpress LLC Alpha-Vitamin E-d6 | 113892-08-3 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN7148S1MG
Encompass_Preferred
Alpha-Vitamin E-d6 is a deuterium-labeled form of alpha-Vitamin E, intended for research use only. Deuterium substitution can influence the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled alpha-Vitamin E
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-18125